Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Serum Rabbit Polyclonal Antibody [clone: 2G9]

Supplier: Rockland Immunochemicals
109-4101 109-4101
RL109-4101EA 319 USD InStock
RL109-4101 CA109-4101
Anti-Serum Rabbit Polyclonal Antibody [clone: 2G9]
Antibodies
Anti-human serum antibody is suitable for ELISA and western blotting. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum and human Serum.

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty