Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FOLR1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
200-103-148S 200-103-148S
RL200-103-148SEA 145 USD InStock
RL200-103-148S CA10036-762
Anti-FOLR1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Anti-Folate-binding Protein has been assayed against 1.0 µg of Folate Binding Protein (Bovine Milk) in a standard capture ELISA using ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:1000 - 1:2500; Western Blot: 1:500 - 1:1000; Immunoprecipitation: 1:100;
Do not add Sodium Azide!

Type: Primary
Antigen: FOLR1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty