Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: 4F3.E8]

Supplier: Rockland Immunochemicals
200-301-F74
RL200-301-F74EA 532 USD InStock
RL200-301-F74
Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: 4F3.E8]
Antibodies
Anti-Hsp90total Antibody is recommended for use in WB, IP, ELISA, Immunofluorescence microscopy and IHC. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:2000

Type: Primary
Antigen: HSP90AA1
Clonality: Monoclonal
Clone: 4F3.E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty