Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CALCA Mouse Monoclonal Antibody [clone: 18C4.B4.C2]

Supplier: Rockland Immunochemicals
200-301-I40 200-301-I40
RL200-301-I40EA 528 USD InStock
RL200-301-I40 CA200-301-I40
Anti-CALCA Mouse Monoclonal Antibody [clone: 18C4.B4.C2]
Antibodies
Anti-Procalcitonin antibody is suitbale for use in ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:1000-1:20000; Western Blot: 1µg/ml;

Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 18C4.B4.C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty