Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCL2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-222 200-401-222
RL200-401-222EA 518 USD InStock
CA200-401-222 RL200-401-222
Anti-BCL2 Rabbit Polyclonal Antibody
Antibodies
This purified IgG antibody against Human Bcl-2 has been tested for use in immunofluorescence microscopy and immunoblotting.  The antibody recognizes a 27 kDa band corresponding to human bcl-2.  Anti-Bcl-2 shows perinuclear staining of Bcl-2.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000;

Type: Primary
Antigen: BCL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty