Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ORF2 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-P11 200-901-P11
RL200-901-P11EA 518 USD InStock
CA200-901-P11 RL200-901-P11
Anti-ORF2 Chicken Polyclonal Antibody
Antibodies
L1/ORF2 Antibody has been tested for use in ELISA.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 149.5 kDa in size corresponding to L1 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:20000-1:100000;

Type: Primary
Antigen: ORF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty