Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD40L Armenian Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MR1]

Supplier: Rockland Immunochemicals
200-B08-N91 200-B08-N91
RL200-B08-N91EA 595 USD InStock
RL200-B08-N91 CA200-B08-N91
Anti-CD40L Armenian Hamster Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MR1]
Antibodies
Anti-CD154 is useful for immunochemistry and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: MR1
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty