Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Sarcosine Oxidase Goat Polyclonal Antibody

Supplier: Rockland Immunochemicals
200101093-0100 200-101-093-0100
RL200101093010EA 518 USD InStock
CARL200101093 RL200101093010
Anti-Sarcosine Oxidase Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Sarcosine Oxidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Microbial

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty