Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PGMB Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-106-237S 200-106-237S
RL200106237SEA 148 USD InStock
RL200106237S CA11029-182
Anti-PGMB Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: PGMB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Lactococcus lacti

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty