Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDCD4 Goat Polyclonal Antibody [clone: 9G6] (Biotin)

Supplier: Rockland Immunochemicals
200-306-964
RL200306964EA 389 USD InStock
RL200306964
Anti-PDCD4 Goat Polyclonal Antibody [clone: 9G6] (Biotin)
Antibodies
Type: Secondary
Antigen: PDCD4
Clonality: Polyclonal
Clone: 9G6
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty