Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FLAG® conjugated Proteins Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-383S
RL600-401-383SEA 145 USD InStock
RL600-401-383S
Anti-FLAG® conjugated Proteins Rabbit Polyclonal Antibody
Antibodies
This antibody is optimally suited for monitoring the expression of FLAG® tagged fusion Proteins. As such, this antibody can be used to identify fusion Proteins containing the FLAG® epitope.

Recommended Dilutions: ELISA: 1:90,000 - 1:250000; Western Blot: 1:2000 - 1:10000;

Type: Tag
Antigen: FLAG® conjugated Proteins
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Carboxy , amino terminal linked FLAG® tagged recombinant Proteins

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty