Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MYC Rabbit Polyclonal Antibody (DyLight® 680)

Supplier: Rockland Immunochemicals
600-444-381 600-444-381
RL600-444-381EA 467 USD InStock
RL600-444-381 CA600-444-381
Anti-MYC Rabbit Polyclonal Antibody (DyLight® 680)
Antibodies
Primary Rabbit Anti-MYC EPITOPE TAG Reacts with Pan

Type: Tag
Antigen: MYC
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 680
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty