Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CFAP206 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
612-401-E42
RL612-401-E42EA 518 USD InStock
RL612-401-E42
Anti-CFAP206 Rabbit Polyclonal Antibody
Antibodies
Anti-Tau pS416 (Rabbit) antibody is suitable for use in Western Blotting. Expect bands or approximately 59, 65, 68 kDa in size corresponding to tau Proteins phosphorylated at Ser416 in the appropriate cell lysate or extract.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CFAP206
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty