Would you like to log in again?
PfuUltra II Fusion HS DNA Polymerase, Agilent Technologies
Supplier: Agilent
600850 600674 600670 600670 600852 600672 600852 600672 600850 600674
ST600672EA 620.6 USD InStock
CA97066-512 ST600672 ST600674 97066-510 CA97066-510 97066-512 99900-198 CAST600674 CA99900-198 CAST600672
PfuUltra II Fusion HS DNA Polymerase, Agilent Technologies
Nucleic Acid Reagents End-point PCR Enzymes and Kits
This DNA polymerase is ideal for a variety of techniques requiring high-fidelity DNA synthesis by the polymerase chain reaction (PCR).
  • Highest fidelity (1 error/2.5 million bp) for the most reliable, error-free PCR
  • Fast extension for 70 to 80% quicker time to results
  • Accurately amplifies up to 19 kB genomic DNA targets
  • Less than 0.5% errors in full length 500 bp PCR products
In Agilent's PfuUltra II Fusion HS DNA Polymerase, we couple the fusion polymerase technology with our engineered PfuUltra DNA Polymerase, hotstart antibody, and proprietary ArchaeMaxx PCR enhancing factor to achieve extreme accuracy, high specificity, and long target-length capability while dramatically reducing overall PCR extension times.
PfuUltra II is the industry standard for high-fidelity PCR. PfuUltra II features a genetically engineered mutant of Pfu DNA polymerase, the ArchaeMaxx Polymerase-Enhancing Factor, fusion technology, and hot start formulation.
PfuUltra II Hotstart PCR Master Mix is a 2X formulation of PfuUltra II Fusion HS DNA Polymerase, an optimized PCR reaction buffer, magnesium, and dNTPs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty