Synonyms:
4-Acetamido-2,2,6,6-Tetramethyl-N-Piperidinyloxy
, 4-Acetamido-2,2,6,6-tetramethylpiperidine 1-Oxyl
, 4-Acetamido-2,2,6,6-tetramethylpiperidine 1-oxyl
A1348-25G
A1348-5G
TCA1348-005GEA
51.23
USD
InStock
TCA1348-005G
TCA1348-25G
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-acetamido-TEMPO) ≥98.0% free radical
4-Acetamido-2,2,6,6-tetramethylpiperidinooxy (4-Acetamido-TEMPO, free radical)
[Catalyst for Oxidation]
CAS Number: 14691-89-5
MDL Number: MFCD00043593
Molecular Formula: C11H21N2O2
Molecular Weight: 213.30
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 143
Flash Point (°C): 198
Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Acetamido-TEMPO