Synonyms:
N-tert-Butyl-alpha-(4-pyridyl)nitrone N'-OxidePOBNN-(4-Pyridylmethylene)-tert-butylamine N,N'-Dioxidealpha-(4-Pyridyl N-Oxide)-N-tert-butylnitrone
B1735-1G
B1735-5G
TCB1735-001GEA
227.26
USD
InStock
TCB1735-001G
TCB1735-5G
N-tert-Butyl-ɑ-(4-pyridyl-1-oxide)nitrone ≥98.0% (by HPLC, titration analysis)
N-tert-Butyl-alpha-(4-pyridyl-1-oxide)nitrone
CAS Number: 66893-81-0
MDL Number: MFCD00041022
Molecular Formula: C10H14N2O2
Molecular Weight: 194.23
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 185
Storage Temperature: 0-10°C
Product Categories: Analytical Science, Spin Trapping Reagents
Other Product Names / Synonyms: N-tert-Butyl-alpha-(4-pyridyl)nitrone N'-Oxide, POBN, N-(4-Pyridylmethylene)-tert-butylamine N,N'-Dioxide, alpha-(4-Pyridyl N-Oxide)-N-tert-butylnitrone