Synonyms:
trans-Cinnamic acid
C0353-500G
C0353-100G
C0353-25G
TCC0353-500GEA
81.5
USD
InStock
TCC0353-500G
TCC0353-025G
TCC0353-100G
trans-Cinnamic acid ≥98.0% (by GC, titration analysis)
trans-Cinnamic acid
CAS Number: 140-10-3
MDL Number: MFCD00004369
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 300
Melting point (°C): 135
Lambda max.: 273 nm (MeOH)
Product Categories: Synthetic Organic Chemistry, Carboxylic Acids (Multi-component Reaction), Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Phenylpropanoids, Phenylpropanoids (Phytochemicals), Analytical Science, Standards for Mass Spectrometry &
Other Product Names / Synonyms: trans-3-Phenylacrylic Acid