Synonyms:
2',7'-Dichloro-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one
D0371-25G
D0371-1G
TCD0371-025GEA
167.18
USD
InStock
TCD0371-025G
TCD0371-001G
2',7'-Dichlorofluorescein ≥95.0% (by HPLC)
2',7'-Dichlorofluorescein
[for Fluorescent indicator]
CAS Number: 76-54-0
MDL Number: MFCD00005047
Molecular Formula: C20H10Cl2O5
Molecular Weight: 401.20
Purity/Analysis Method: 95.0% (HPLC)
Form: Crystal
Melting point (°C): 280