Synonyms:
2-Chloro-n,n-diethylacetamide
, N,N,-Diethylchloroacetamide
D0483-25G
D0483-500G
TCD0483-25GEA
43.59
USD
InStock
TCD0483-25G
TCD0483-500G
2-Chloro-N,N-diethylacetamide ≥98.0%
2-Chloro-N,N-diethylacetamide
CAS Number: 2315-36-8
MDL Number: MFCD00000928
Molecular Formula: C6H12ClNO
Molecular Weight: 149.62
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 110
Specific Gravity (20/20): 1.09
Other Product Names / Synonyms: N-Chloroacetyldiethylamine