Synonyms:
(1-Pyridinio)acetohydrazide chloride
, 1-(2-Hydrazinyl-2-oxoethyl)pyridin-1-ium chloride
, 1-(Hydrazinocarbonylmethyl)pyridinium chloride
, Girard's Reagent P
G0030-25G
TCG0030-025GEA
185.27
USD
InStock
TCG0030-025G
Girard's reagent P (carbazoylmethylpyridinium chloride) ≥95.0% (by titrimetric analysis)
Girard's reagent P (Carbazoylmethylpyridinium chloride)
CAS Number: 1126-58-5
MDL Number: MFCD00011992
Molecular Formula: C7H10ClN3O
Molecular Weight: 187.63
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 203
Product Categories: Analytical Science, Derivatization Reagents for MS
Other Product Names / Synonyms: 1-(Hydrazinocarbonylmethyl)pyridinium Chloride, (1-Pyridinio)acetohydrazide Chloride