Synonyms:
Glutaral 1,5-Pentanedial
G0068-500ML
G0068-25ML
TCG0068-25MLEA
23.7
USD
InStock
TCG0068-25ML
TCG0068-500ML
Glutaraldehyde ∼50% ∼5.6 mol/L in water
Glutaraldehyde
CAS Number: 111-30-8
MDL Number: MFCD00007025
Molecular Formula: C5H8O2
Molecular Weight: 100.12
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.13