Synonyms:
HMDSO Dow Corning 200® fluid/0.65 cSt Silicone fluid DC 200/0.65 cSt
H0091-100ML
H0091-25ML
H0091-500ML
TCH0091-500MLEA
59.06
USD
InStock
TCH0091-500ML
TCH0091-25ML
TCH0091-100ML
Hexamethyldisiloxane, 0.65 cSt (25°C) ≥98.0%
Hexamethyldisiloxane
CAS Number: 107-46-0
MDL Number: MFCD00008265
Molecular Formula: C6H18OSi2
Molecular Weight: 162.38
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): -2
Specific Gravity (20/20): 0.77