Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Hydroxyphenylazo)benzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: 4'-Hydroxyazobenzene-2-carboxylic acid2-((4-Hydroxyphenyl)diazenyl)benzoic acidHABA2-(4-Hydroxyphenylazo)benzoic acid4-Hydroxyazobenzene-2'-carboxylic acid

H0586-25G H0586-5G
TCH0586-005GEA 64.48 USD InStock
TCH0586-005G TCH0586-25G
2-(4-Hydroxyphenylazo)benzoic acid ≥98.0% (by HPLC, titration analysis)
2-(4-Hydroxyphenylazo)benzoic acid
CAS Number: 1634-82-8
MDL Number: MFCD00002428
Molecular Formula: C13H10N2O3
Molecular Weight: 242.23
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 201

Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: HABA, 2-(4-Hydroxyphenylazo)benzoic Acid, 4-Hydroxyazobenzene-2'-carboxylic Acid


Formula: C₁₃H₁₀N₂O₃
MW: 242.23 g/mol
Melting Pt: 201 °C
MDL Number: MFCD00002428
CAS Number: 1634-82-8
EINECS: 216-655-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/98/28970598.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty