Synonyms:
3-Hydroxypyridine-2-carboxylic acid
H0787-5G
H0787-25G
TCH0787-005GEA
94.4
USD
InStock
TCH0787-005G
TCH0787-25G
3-Hydroxypicolinic acid ≥98.0% (by titrimetric analysis)
3-Hydroxypicolinic acid
CAS Number: 874-24-8
MDL Number: MFCD00006294
Molecular Formula: C6H5NO3
Molecular Weight: 139.11
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 204
Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 3-Hydroxypicolinic Acid