Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) ≥98.0% free radical

Supplier: TCI America
Synonyms: 2,2,6,6-Tetramethyl-4-hydroxy-1-piperidinyloxy radicalTempol4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-Oxyl

H0865-25G H0865-5G
TCH0865-025GEA 212.06 USD InStock
TCH0865-025G TCH0865-005G
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-hydroxy-TEMPO, TEMPOL) ≥98.0% free radical
4-Hydroxy-2,2,6,6-tetramethylpiperidinooxy (4-Hydroxy-TEMPO, TEMPOL, free radical)
CAS Number: 2226-96-2
MDL Number: MFCD00006478
Molecular Formula: C9H18NO2
Molecular Weight: 172.25
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 72
Flash Point (°C): 146

Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Hydroxy-TEMPO


Formula: C₉H₁₈NO₂
MW: 172.25 g/mol
Melting Pt: 72 °C
Flash Pt: 146 °C
MDL Number: MFCD00006478
CAS Number: 2226-96-2
EINECS: 218-760-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/91/28970891.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty