Synonyms:
4-Hydroxy-TEMPO Benzoate4-Benzoyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl4-Benzoyloxy-TEMPO
H0878-1G
H0878-5G
TCH0878-005GEA
244.62
USD
InStock
TCH0878-005G
TCH0878-001G
4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate ≥97.0% free radical
4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate
[Catalyst for Oxidation]
CAS Number: 3225-26-1
MDL Number: MFCD00075563
Molecular Formula: C16H22NO3
Molecular Weight: 276.36
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 104
Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Hydroxy-TEMPO Benzoate, 4-Benzoyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Benzoyloxy-TEMPO