Synonyms:
2-Hydrazinopyridine2-hydrazinylpyridine2(1H)-Pyridinone, hydrazone
H0888-10G
H0888-25G
TCH0888-010GEA
163.85
USD
InStock
TCH0888-010G
TCH0888-25G
2-Hydrazinylpyridine ≥98.0% (by GC, titration analysis)
2-Hydrazinylpyridine
CAS Number: 4930-98-7
MDL Number: MFCD00006249
Molecular Formula: C5H7N3
Molecular Weight: 109.13
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 47
Flash Point (°C): 110
Product Categories: Analytical Science, Derivatization Reagents for MS
Other Product Names / Synonyms: 2-Pyridylhydrazine