Synonyms:
3-(3-Indolyl)acrylic AcidIAA
I0025-1G
I0025-5G
TCI0025-001GEA
66.99
USD
InStock
TCI0025-001G
TCI0025-005G
3-Indoleacrylic acid ≥98.0% (by HPLC, titration analysis)
3-Indoleacrylic acid
CAS Number: 1204-06-4
MDL Number: MFCD00005633
Molecular Formula: C11H9NO2
Molecular Weight: 187.20
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 3-(3-Indolyl)acrylic Acid, IAA