Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Isothiocyanato-2,2,6,6-tetramethylpiperidine 1-oxyl ≥97.0% (by GC, titration analysis) free radical

Supplier: TCI America
Synonyms: 4-Isothiocyanato-TEMPO

I0486-100MG I0486-1G
TCI0486-1GEA 942.39 USD InStock
TCI0486-1G TCI0486-100MG
4-Isothiocyanato-2,2,6,6-tetramethylpiperidine 1-oxyl ≥97.0% (by GC, titration analysis) free radical
4-Isothiocyanato-2,2,6,6-tetramethylpiperidine 1-oxyl
CAS Number: 36410-81-8
MDL Number: MFCD00010109
Molecular Formula: C10H17N2OS
Molecular Weight: 213.32
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Melting point (°C): 128
Storage Temperature: 0°C

Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Isothiocyanato-TEMPO


Formula: C₁₀H₁₇N₂OS
MW: 213.32 g/mol
Melting Pt: 128 °C
MDL Number: MFCD00010109
CAS Number: 36410-81-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/23/42/28972342.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty