N0367-25G
N0367-5G
TCN0367-025GEA
225
USD
InStock
TCN0367-025G
TCN0367-5G
Nitrosobenzene ≥98.0%
Nitrosobenzene
CAS Number: 586-96-9
MDL Number: MFCD00002059
Molecular Formula: C6H5NO
Molecular Weight: 107.11
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 57
Melting point (°C): 66
Storage Temperature: 0-10°C