Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Nitrobenzyl alcohol ≥99.0%

Supplier: TCI America
Synonyms: (3-Nitrophenyl)methanol

S0378-1G S0378-10G
TCS0378-1GEA 28 USD InStock
TCS0378-1G TCS0378-10G
3-Nitrobenzyl alcohol ≥99.0%
3-Nitrobenzyl alcohol
[Matrix for FABMS and liquid SIMS]
CAS Number: 619-25-0
MDL Number: MFCD00007273
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Flash Point (°C): 110
Freezing point (°C): 30


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00007273
CAS Number: 619-25-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/40/91/28994091.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty