Synonyms:
(3-Nitrophenyl)methanol
S0378-1G
S0378-10G
TCS0378-1GEA
28
USD
InStock
TCS0378-1G
TCS0378-10G
3-Nitrobenzyl alcohol ≥99.0%
3-Nitrobenzyl alcohol
[Matrix for FABMS and liquid SIMS]
CAS Number: 619-25-0
MDL Number: MFCD00007273
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Flash Point (°C): 110
Freezing point (°C): 30