Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium [(S)-1,2-diaminopropane-N,N,N',N'-tetraacetato]samarate(III) ≥90.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Sodium [(S)-Propylenediamine-N,N,N',N'-tetraacetato]samarate(III)

S0474-100MG
TCS0474-100MGEA 538.74 USD InStock
TCS0474-100MG
Sodium [(S)-1,2-diaminopropane-N,N,N',N'-tetraacetato]samarate(III) ≥90.0% (by titrimetric analysis)
Sodium [(S)-1,2-diaminopropane-N,N,N',N'-tetraacetato]samarate(III)
MDL Number: MFCD06797165
Molecular Formula: C11H15N2O8Sm
Molecular Weight: 475.59
Purity/Analysis Method: 90.0% (T)
Form: Crystal
Color: White
Specific rotation [a]20/D: 5 deg (C=1, pH 10.0 Buffer sol.)

Other Product Names / Synonyms: Sodium [(S)-Propylenediamine-N,N,N',N'-tetraacetato]samarate(III)


https://us-legacy.vwr.com/stibo/web/std.lang.all/41/34/28994134.jpg
Order Now
Specification Test Results
Purity/Analysis Method 90.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty