Synonyms:
N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochloride
, N1,N1,N4,N4-Tetramethylbenzene-1,4-diamine dihydrochloride
, TMPD 2 HCl
, N,N,N',N'-Tetramethyl-1,4-phenylenediammonium dichloride
T0151-5G
T0151-1G
T0151-25G
TCT0151-5GEA
47.71
USD
InStock
TCT0151-5G
TCT0151-001G
TCT0151-025G
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen)
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
CAS Number: 637-01-4
MDL Number: MFCD00012482
Molecular Formula: C10H16N2
Molecular Weight: 237.17
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 224
Other Product Names / Synonyms: TMPD