Synonyms:
TMS
T0154-25ML
T0154-500ML
T0154-100ML
TCT0154-100MLEA
119.74
USD
InStock
TCT0154-100ML
TCT0154-25ML
TCT0154-500ML
Tetramethylsilane ≥99.0%
Tetramethylsilane
[for NMR]
CAS Number: 75-76-3
MDL Number: MFCD00008274
Molecular Formula: C4H12Si
Molecular Weight: 88.23
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 26
Flash Point (°C): -27
Specific Gravity (20/20): 0.64
Storage Temperature: 0-10°C