Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-α-Terpineol ≥95.0%

Supplier: TCI America
Warning
Synonyms: 2-(4-Methylcyclohex-3-en-1-yl)propan-2-olTerpineolα-Terpineol

T0984-100ML T0984-500ML T0984-25ML
TCT0984-500MLEA 289.28 USD InStock
TCT0984-500ML TCT0984-025ML TCT0984-100ML
(±)-α-Terpineol ≥95.0%
(±)-α-Terpineol
CAS Number: 98-55-5
MDL Number: MFCD00001557
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 218
Flash Point (°C): 101
Freezing point (°C): 31

Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: 1-p-Menthen-8-ol


Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 218 °C
Flash Pt: 101 °C
MDL Number: MFCD00001557
CAS Number: 98-55-5
EINECS: 202-680-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/50/68/28995068.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty