Synonyms:
3-(Trimethylsilyl)-1-propanesulfonic Acid Sodium SaltDSSSodium 2,2-Dimethyl-2-silapentane-5-sulfonate
T1638-1G
T1638-5G
TCT1638-1GEA
256.38
USD
InStock
TCT1638-1G
TCT1638-005G
Sodium-3-(trimethylsilyl)-1-propanesulfonate ≥98.0% (by titrimetric analysis)
Sodium-3-(trimethylsilyl)-1-propanesulfonate
[1H NMR Standard for D2O Solvent]
CAS Number: 2039-96-5
MDL Number: MFCD00007537
Molecular Formula: C6H16O3SSi
Molecular Weight: 218.32
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Product Categories: Analytical Science, Standards for NMR
Other Product Names / Synonyms: 3-(Trimethylsilyl)-1-propanesulfonic Acid Sodium Salt, DSS, Sodium 2,2-Dimethyl-2-silapentane-5-sulfonate