Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
VAI® SMA MicroParticle ICS, Veltek Associates
Supplier: Veltek
SMA-MP-PS SMA-MP-WLB SMA-MP-PAP SMA-MP-WM SMA-MP-TA SMA-MP-IOQ SMA-MP-CH SMA-MP-HC SMA-MP-HH SMA-MP-USB SMA-MP-PUK SMA-MP-PNA SMA-MP-PCE SMA-MP-BTY SMA-MP-PTR SMA-MP-PRA SMA-MP-BA SMA-MP-PAU SMA-MP-TB SMA-MP-TRH
76326-580EA 78.41 USD InStock
76326-580 76326-550 76326-582 76326-576 76326-510 76326-520 76326-542 76326-512 76326-578 76326-522 76326-544 76326-514 76326-546 76326-524 76326-516 76326-548 76326-504 76326-518 76326-506 76326-508
VAI® SMA MicroParticle ICS, Veltek Associates
Particle Counters
As part of the Environmental Contol Monitoring Division, the SMA MicroParticle ICS systems are used for continuous or periodic monitoring of particle counts in cleanrooms and critical environments.
  • Multi-processing allows the instrument to simultaneously process, perform tasks, and log data such as humidity, temperature, and particle counts at full speed without interrupting sampling
  • Real-time meter displays the number of particles per second, per channel, which assists users at pinpointing the exact location of contamination sources
  • Sample time can be configured from 1 second to 99 hours with sample management features allowing 1000 user defined sampling locations with up to 50 recipes
  • Annotations allow users to add note fields to any data record while the instrument is sampling
  • Units have advanced power management features including sleep mode, and over 10 hours of battery life
  • Units can store up to 45000 comprehensive data records for each sample
  • Units offer user definable reports, come with internal audible alarms, and are easy to clean and wipe down with minimal particle traps
All particle counter models are equipped with a 7.6 cm (3") display screen. The Handheld model features a lightweight, high-impact injection molded plastic enclosure. Both the Table-top and Wall Mounted models feature lightweight, stainless steel enclosures.
Certifications: Units can print or display reports that comply to ISO 14644-1, EU GMP Annex 1, and federal standard 209E. Complies with ISO 21501-4 and JIS B9921 standards.
Accessories information: Included and replacement accessories come standard with purchase of unit (exception is the rechargeable battery; rechargeable battery is only included with Handheld and Table-top models). Optional accessories are not included.
Ordering information: All units are supplied with a one-year warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty