Would you like to log in again?
Battery Powered Milliohm Meter, Extech
Supplier: Extech
380580-NIST 380580
89370-126EA 1598.15 USD InStock
89370-126 10171-814
Battery Powered Milliohm Meter, Extech
Electrical Meters Digital Ohmmeters
High accuracy, high resolution portable Milliohm Meter for field use with 5 ranges with 100µOhm max resolution
This meter provides over-temperature and over-voltage protection as well as Invalid test indicators prevent inaccurate measurements.
Certifications: CE marked.
Ordering information: Comes with 8AA batteries, and 2 sets of test leads (4 wire with 4 alligator clips and 4 wire with 2 Kelvin alligator clips), built-in water resistant case with shoulder strap. 1-Year Warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty