Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antimycin A (from Streptomyces species)

Supplier: Thermo Scientific Chemicals
Danger

J63522-MA J63522-LB0 J63522-MA
AAJ63522-MAEA 140.01 USD InStock
AAJ63522-MA CAAAJ63522-MA AAJ63522-LB0
Antimycin A (from Streptomyces species)
Antimycin A
Antimycin A is used to study the specific sites of reactive oxygen species production in mitochondria isolated from skeletal muscle of chronic obstructive pulmonary disease patients, and its relationship with local oxidative stress induced by exercise.

  • Light sensitive

White to pale yellow powder

Soluble in 100% ethanol and insoluble in water

Caution: Research use only: Not intended for animal or human diagnostic or therapeutic use.


Formula: C₂₈H₄₀N₂O₉
Melting Pt: 149…150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01779723
CAS Number: 1397-94-0
UN: 3462
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty