Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Heavy Duty Vibration Meter, Extech
Supplier: Extech
407860 407860-NIST
10171-754EA 1794.51 USD InStock
10171-754 97050-006
Heavy Duty Vibration Meter, Extech
Vibration Meters
Heavy Duty Vibration Meter that measures Acceleration, Velocity, and Displacement with a built-in PC interface
This meter features a Record mode that stores the maximum and minimum values for later recall. Other features include RMS or Peak Value measurement modes and Data Hold, Auto shut-off functions. Powered by 9V battery power or optional AC adaptor.
Certifications: CE marked.
Ordering information: Remote sensor, magnetic mount, 9V battery, protective rubber holster with stand, and hard carrying case included. 1 year warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty