Would you like to log in again?
Electromagnetic Coil, Horseshoe Core Design, Ace Glass
Supplier: Ace Glass
6593-40
89055-680EA 984.4 USD InStock
89055-680
Electromagnetic Coil, Horseshoe Core Design, Ace Glass
Magnets
This electromagnetic coil is a potted type “horseshoe” core design used to operate large automatic distilling heads, up to 31/4 inch (83 mm) O.D
with adjustable clamp. Operates at 120 volts, 50/60 Hz AC maximum, delivering 24 watts of pulling power. For 100% duty cycle operation.
Ordering information: The unit is supplied with a 6-foot, 3-prong cord and a grounded aluminum cover. Fabricated from borosilicate glass. Comes with a one-time 90-day unconditional warranty. Contact your Avantor representative for details.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty