Would you like to log in again?
IntelliCAL™ PHC201 Standard Gel Filled pH Electrode, 1 m cable, Hach
Supplier: Hach
PHC20101 PHC20101
89347-984EA 647.68 USD InStock
89347-984 CA11028-182
IntelliCAL™ PHC201 Standard Gel Filled pH Electrode, 1 m cable, Hach
Electrodes pH Electrodes
IntelliCAL™ PHC201 is a digital, combination pH probe with a non-refillable, gel-filled double junction reference and built-in temperature sensor.
  • IntelliCAL™ PHC201 provides fast, stable response in water quality applications.
  • IntelliCAL™ digital probes provide ultimate traceability in measurement history.
  • IntelliCAL™ digital probes alert the user when recalibration is needed.
  • IntelliCAL™ digital probes can be moved between meters without the need to recalibrate or reenter measurement settings.
It is available with a 1 or 3 meter cable and is intended for laboratory use. The PHC201 has a ceramic pin reference junction and is ideal for measuring pH in general/clean water quality applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty