Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EMF Strength Meter, 8 GHz RF, Extech

Supplier: Extech
480846
89370-136EA 1337.49 USD InStock
89370-136
EMF Strength Meter, 8 GHz RF, Extech
Electromagnetic Field Meters
For electromagnetic field strength measurement including mobile phone base station antenna radiation, RF power measurement for transmitters, WAN detection/installation, wireless communication applications (CW, TDMA, GSM, DECT) and microwave leakage

Complete with 9V battery and carrying case.

Audible alarm with user selectable threshold.

Certifications: CE marked.

Ordering information: One-year warranty.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty