Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.

EMF Strength Meter, 3.5 GHz RF, Extech

Supplier: Extech
480836
89370-138EA 575.33 USD InStock
89370-138
EMF Strength Meter, 3.5 GHz RF, Extech
Electromagnetic Field Meters
For electromagnetic field strength measurement of mobile phones, cell phone and base stations, microwave leakage

Complete with 9V battery and carrying case.

It has a user selectable alarm threshold.

Certifications: CE marked.

Ordering information: One-year warranty.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty