Would you like to log in again?
Triple Axis EMF Tester, Extech
Supplier: Extech
480826
89370-140EA 629.68 USD InStock
89370-140
Triple Axis EMF Tester, Extech
Electromagnetic Field Meters
Designed to provide user a quick, reliable and easy way to measure EMF radiation levels around power lines, electrical appliances and industrial devices
Complete with sensor with 3ft. (930mm) cable, 9V battery, and hard carrying case.
Big digit LCD display with data hold and easy to use remote probe.
Certifications: CE marked.
Ordering information: One-year warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty