Would you like to log in again?
EMF/ELF Meter, Extech
Supplier: Extech
480823
89370-142EA 224.32 USD InStock
89370-142
EMF/ELF Meter, Extech
Measures electromagnetic field radiation levels from fans, electrical appliances, wiring, and power lines
Complete with 9V battery.
The meter has a large 1/2" LCD (1999 count) display of Electromagnetic Field (EMF) level in milliGauss or microTesla.
Certifications: CE marked.
Ordering information: One-year warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty