Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.

EZ40 EzFlex™ Combustible Gas Detector, Extech

Supplier: Extech
EzFlex™
EZ40
89370-184EA 397.59 USD InStock
89370-184
EZ40 EzFlex™ Combustible Gas Detector, Extech
Leak Detection Systems Gas Leak Detection Systems
The EZ40 EzFlex™ Combustible Gas Detector quickly identifies and pinpoints gas leaks

Three "C" batteries are included.

The unit features one-hand operation with thumb controlled sensitivity adjustments to eliminate background gas levels. It offers high sensitivity and is able to locate the smallest leaks. It also features a visible and audible alarm at 10% (LEL) Lower Explosive Limity for methane.

Pump driven field calibration range: 10 ppm

Warm-up time: Approx. 1 Minute

Certifications: CE marked.

Ordering information: Unit is supplied with a 1-year warranty.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty