Would you like to log in again?
CG204 Coating Thickness Tester, Extech
Supplier: Extech
CG204
89200-736EA 546 USD InStock
89200-736
CG204 Coating Thickness Tester, Extech
Coating Testers
Coating Thickness Tester with automatic recognition of ferrous and non-ferrous substrates
The CG204 features an easy-to use menu system with two measuring modes (Single and Continuous) and two working modes (Direct and Group). The memory stores 400 readings: 80 Direct and 320 Group. Other features include user-programmable high/low alarms, Min/Max/Average, and one or two point calibration.
Resolution: 0.1 μm (0.004 mils)
Working principle of magnetic induction (ferrous); Eddy current (non-ferrous).
Certifications: CE
Ordering information: Complete with two AAA batteries, USB cable, software, Calibration Iron, Calibration Aluminum, Precision Standards, and pouch case.
1 yr warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty