Would you like to log in again?
USB Sound Level Data Logger, Extech
Supplier: Flir
407760 407760-NISTL
10171-758EA 881.96 USD InStock
10171-758 97050-010
USB Sound Level Data Logger, Extech
Sound Meters
High accuracy sound-level datalogging of up to 129,920 records
Model 407760 features USB interface for easy setup and data download and Records readings with real time clock. Two start methods: programmed (from PC) or manual. Other features include a 0.5" microphone. A and C weighting.
Certifications: CE marked.
Ordering information: Complete with 3.6V Lithium battery, Windows® compatible software, USB cover, tripod, USB extension cable, and windscreen. One year warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty